Skip to Content
Merck
All Photos(1)

Key Documents

239763

Sigma-Aldrich

1-Decanol

≥98%

Synonym(s):

n-Decyl alcohol, Alcohol C10

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$100.00

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$100.00

About This Item

Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
Beilstein:
1735221
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

4.5 (vs air)

Quality Level

vapor pressure

1 mmHg ( 70 °C)
8.25 mmHg ( 100 °C)

Assay

≥98%

form

liquid

autoignition temp.

550 °F

refractive index

n20/D 1.437 (lit.)

bp

231 °C (lit.)

mp

5-7 °C (lit.)

solubility

alcohol: soluble(lit.)
diethyl ether: soluble(lit.)
water: insoluble(lit.)

density

0.829 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCO

InChI

1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3

InChI key

MWKFXSUHUHTGQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The interaction of 1-decanol with nanosilicas and silica gels were analyzed using differential scanning calorimetry (DSC) and thermoporometry[1].

Application

1-Decanol was used as an extraction solvent in the determination of several toxic phenolic compounds present in cigarette smoke by single-drop microextraction in combination with liquid chromatography-tandem mass spectrometry[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - Pensky-Martens closed cup

Flash Point(C)

95 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anika Reinhardt et al.
Stem cell reports, 14(3), 520-527 (2020-02-29)
A common strategy for multi-protein expression is to link genes by self-cleaving 2A peptide sequences. Yet, little is known how the 2A peptide-derived N-terminal proline or adjacent non-native residues introduced during cDNA cloning affects protein stoichiometry. Polycistronic reprogramming constructs with
Subhrakanti Saha et al.
Analytical and bioanalytical chemistry, 405(28), 9265-9272 (2013-09-24)
Cigarette smoke contains several toxic phenolic compounds, measurements of which are essential from a public health standpoint. This article describes a simple and selective analytical method for quantitative determination of six toxic phenolic compounds (phenol, catechol, resorcinol, hydroquinone, o-cresol, and
Prabakaran D S et al.
PloS one, 17(4), e0266532-e0266532 (2022-04-15)
Human papillomavirus type 16 (HPV16) plays a major role in the development of cervical cancer. The oncogenic potential of HPV16 is attributed to E6 and E7 oncoproteins. Here, we investigated the relationship between fused toes homolog (FTS) and HPV16 E6
Vladimir V Turov et al.
Langmuir : the ACS journal of surfaces and colloids, 29(13), 4303-4314 (2013-03-14)
The interfacial and temperature behavior of n-decane bound to weakly hydrated nanosilica A-400 (initial, heated, or compacted) or silica gel Si-60 was studied using low-temperature (1)H NMR spectroscopy applied to static samples that allowed us to observe signals only of
Shihu Gan et al.
RNA biology, 18(6), 900-913 (2020-10-16)
The mechanisms that regulate cell-cycle arrest of cardiomyocytes during heart development are largely unknown. We have previously identified Tudor staphylococcal nuclease (Tudor-SN) as a cell-cycle regulator and have shown that its expression level was closely related to cell-proliferation capacity. Herein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service