Skip to Content
Merck
All Photos(1)

Key Documents

01977

Supelco

Silica gel 60 ADAMANT on TLC plates

with fluorescent indicator 254 nm

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41115711
PubChem Substance ID:
NACRES:
NC.07

quality

with fluorescent indicator 254 nm

Quality Level

feature

fluorescent indicator

technique(s)

thin layer chromatography (TLC): suitable

layer thickness

250 μm ± 30 μm

plate size

2.5 cm × 7.5 cm

particle size

7.0-11.0 μm

pore size

60 Å medium pore diameter

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Adamant is a trademark of Macherey-Nagel GmbH & Co., KG

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service