Z293032
TLC Glass Plates
L × W 20 cm × 20 cm, C18-silica gel 60 matrix, binder, Polymeric, fluorescent indicator
Synonym(s):
TLC Silica gel 60 RP-18 F254s, TLC Silica gel 60 RP-18 F254s, 25 Glass plates 20 x 20 cm
About This Item
Recommended Products
material
C18-silica gel 60 matrix
glass support
feature
binder Polymeric
fluorescent indicator
packaging
pkg of 25 ea
manufacturer/tradename
EMD Millipore® 1.15389.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
250 μm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Packaging
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service