Skip to Content
Merck
All Photos(1)

Key Documents

00891

Supelco

Dextran

analytical standard, for GPC, 50,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10O5]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

form

powder or crystals

mol wt

Mn ~35,600
Mp ~43,500
Mw ~48,600

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.36

application(s)

food and beverages

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dextran may be used as a reference standard in the separation of dextrans from Leuconostoc mesenteroids B512 using gel permeation chromatography (GPC).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of dextrans by gel permeation chromatography.
Soeteman AA, et al.
Journal of Polymer Science. Part B, Polymer Physics, 16(12), 2147-2155 (1978)
Dmitri Simberg et al.
Biomaterials, 30(23-24), 3926-3933 (2009-04-28)
In order to understand the role of plasma proteins in the rapid liver clearance of dextran-coated superparamagnetic iron oxide (SPIO) in vivo, we analyzed the full repertoire of SPIO-binding blood proteins using novel two-dimensional differential mass spectrometry approach. The identified
Elizabeth M Bradshaw et al.
Nature neuroscience, 16(7), 848-850 (2013-05-28)
In our functional dissection of the CD33 Alzheimer's disease susceptibility locus, we found that the rs3865444(C) risk allele was associated with greater cell surface expression of CD33 in the monocytes (t50 = 10.06, P(joint) = 1.3 × 10(-13)) of young
Yoshikazu Isomura et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(25), 10209-10220 (2013-06-21)
It is widely accepted that dorsal striatum neurons participate in either the direct pathway (expressing dopamine D1 receptors) or the indirect pathway (expressing D2 receptors), controlling voluntary movements in an antagonistically balancing manner. The D1- and D2-expressing neurons are activated
R Shelma et al.
Journal of biomedical nanotechnology, 9(2), 167-176 (2013-05-01)
The overall goal of this paper was to enhance the bioavailability of orally delivered insulin. A mucoadhesive amphiphilic chitosan derivative, lauroyl sulphated chitosan (LSCS) was used as the oral carrier for insulin. Trans epithelial electrical resistance experiment was performed on

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service