Skip to Content
Merck
All Photos(1)

Documents

00896

Supelco

Dextran

analytical standard, for GPC, 670,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10O5]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

form

powder or crystals

mol wt

Mn ~332,800
Mp ~401,300
Mw ~667,800

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~2.01

application(s)

food and beverages

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Dextran in drug delivery: Research highlights the utilization of dextran in formulating hydrogels for enhanced wound healing, where collagen-polyurethane-dextran hydrogels demonstrate significant anti-inflammatory effects and promote collagen synthesis, thus supporting tissue regeneration processes (Aguayo-Morales et al., 2024).

  • Dextran as a tool in bioconjugation: A study investigated the role of dextran in bioconjugation to improve the cellular uptake of antisense oligonucleotides. This application of dextran enhances the delivery mechanisms of nucleic acid therapies, showing promise for genetic disorders and therapeutic interventions (Kohashi et al., 2024).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nataly Puerta Cavanzo et al.
Frontiers in medicine, 7, 615774-615774 (2021-02-02)
An unsolved therapeutic problem in fibrosis is the overproduction of collagen. In order to screen the effect of anti-fibrotic drugs on collagen deposition, the Scar-in-a-Jar approach has been introduced about a decade ago. With macromolecular crowding a rapid deposition of
Dmitri Simberg et al.
Biomaterials, 30(23-24), 3926-3933 (2009-04-28)
In order to understand the role of plasma proteins in the rapid liver clearance of dextran-coated superparamagnetic iron oxide (SPIO) in vivo, we analyzed the full repertoire of SPIO-binding blood proteins using novel two-dimensional differential mass spectrometry approach. The identified
Elizabeth M Bradshaw et al.
Nature neuroscience, 16(7), 848-850 (2013-05-28)
In our functional dissection of the CD33 Alzheimer's disease susceptibility locus, we found that the rs3865444(C) risk allele was associated with greater cell surface expression of CD33 in the monocytes (t50 = 10.06, P(joint) = 1.3 × 10(-13)) of young
R Shelma et al.
Journal of biomedical nanotechnology, 9(2), 167-176 (2013-05-01)
The overall goal of this paper was to enhance the bioavailability of orally delivered insulin. A mucoadhesive amphiphilic chitosan derivative, lauroyl sulphated chitosan (LSCS) was used as the oral carrier for insulin. Trans epithelial electrical resistance experiment was performed on
Editorial comment.
Joseph G Barone
Urology, 81(5), 1045-1046 (2013-04-24)

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service