Skip to Content
Merck
All Photos(1)

Key Documents

1179708

USP

Dextran 40

Dextran 40 United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$666.00

$666.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$666.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$666.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

dextran

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Dextran 40 is primarily composed of glucose units linked together, with the predominant type of linkage being α-1:6. It is obtained through controlled breakdown and separation of certain polysaccharides that are synthesized when certain strains of Leuconostoc mesenteroides bacteria ferment sucrose. Dextran 40 has a weight average molecular weight within the range of 35,000 to 45,000. [1]

Application

Dextran 40 USP reference standard for use in specified quality tests and assays.

Also used to prepare a system suitability solution during optical and specific rotation of Dextran 40 by liquid chromatography method coupled to a Refractive index detector. [1]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dextran 40
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 47(5), 1309-1309 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service