Skip to Content
Merck
All Photos(1)

Key Documents

21-0740

Sigma-Aldrich

N-(1-Naphthyl)ethylenediamine dihydrochloride

≥97.0%, suitable for determination of nitroxide

Synonym(s):

2-(1-Naphthylamino)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
259.17
Beilstein:
3707471
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97.0%

form

powder

availability

available only in Japan

mp

194-198 °C (dec.) (lit.)

suitability

suitable for determination of nitroxide

storage temp.

15-25°C

SMILES string

Cl.Cl.NCCNc1cccc2ccccc12

InChI

1S/C12H14N2.2ClH/c13-8-9-14-12-7-3-5-10-4-1-2-6-11(10)12;;/h1-7,14H,8-9,13H2;2*1H

InChI key

MZNYWPRCVDMOJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigenori Nakano et al.
Talanta, 81(3), 786-791 (2010-03-20)
A novel flow-injection spectrophotometry has been developed for the determination of molybdenum(VI) at nanograms per milliliter levels. The method is based on the catalytic effect of molybdenum(VI) on the bromate oxidative coupling of p-hydrazinobenzenesulfonic acid with N-(1-naphthyl)ethylenediamine to form an
P K Laikind et al.
Analytical biochemistry, 156(1), 81-90 (1986-07-01)
The Bratton-Marshall reaction can be used to identify patients with adenylosuccinate lyase deficiency. These patients excrete in their urine the dephosphorylated derivative of the de novo purine synthesis intermediate 5'-phosphoribosyl-4-(N-succinylcarboxamide)-5-aminoimidazole (SAICAR). The test described here depends on a coupling reaction
Improved dye procedure for determining urea concentration by using o-phthalaldehyde and naphthylethylenediamine.
N T Lequang et al.
Clinical chemistry, 33(1), 192-192 (1987-01-01)
A C Roy et al.
Electrophoresis, 13(6), 396-397 (1992-06-01)
A sensitive staining method for the detection of oxytocinase (EC 3.4.11.3) activity in electrophoresis gels has been described. The method is based on the enzymatic release of p-nitroaniline (PNA) from two specific synthetic oxytocinase substrates, S-benzyl-L-cysteine-p-nitroanilide (BCN) and L-leucine-p-nitroanilide (LN)
J M Fujitaki et al.
Journal of biochemical and biophysical methods, 29(2), 143-148 (1994-09-01)
A simple spectrophotometric assay has been developed for determining concentrations of the new cardioprotective agent, acadesine (AICA-riboside), in human plasma. The method is adapted from the Bratton-Marshall (BMA) procedure for detection of primary aromatic amines. The assay was developed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service