Skip to Content
Merck
All Photos(2)

Key Documents

700001P

Avanti

DC-Cholesterol HCl

Avanti Research - A Croda Brand 700001P, powder

Synonym(s):

3β-[N-(N′,N′-dimethylaminoethane)-carbamoyl]cholesterol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$187.00
200 MG
$509.00
500 MG
$985.00
1 G
$1,640.00

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$187.00
200 MG
$509.00
500 MG
$985.00
1 G
$1,640.00

About This Item

Empirical Formula (Hill Notation):
C32H57N2O2Cl
CAS Number:
Molecular Weight:
537.26
UNSPSC Code:
12352211
NACRES:
NA.25

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (700001P-1g)
pkg of 1 × 200 mg (700001P-200mg)
pkg of 1 × 25 mg (700001P-25mg)
pkg of 1 × 500 mg (700001P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700001P

application(s)

vaccine development

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](C1)(OC(NCC[N+](C)([H])C)=O)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](C)CCCC(C)C.[Cl-]

InChI

1S/C32H56N2O2.ClH/c1-22(2)9-8-10-23(3)27-13-14-28-26-12-11-24-21-25(36-30(35)33-19-20-34(6)7)15-17-31(24,4)29(26)16-18-32(27,28)5;/h11,22-23,25-29H,8-10,12-21H2,1-7H3,(H,33,35);1H

InChI key

ISXSJGHXHUZXNF-UHFFFAOYSA-N

General description

DC-Cholesterol HCl is a cholesterol derivative and a cationic lipid[1] with a head group and linker moiety.[2]
DC-Cholesterol HCl is a sterol-based transfection reagent.

Application

DC-Cholesterol HCl is suitable:
  • for the preparation of ultrasound cationic microbubbles[3]
  • to prepare liposomes[4]
  • for the preparation of L-glutathione (GSH), loaded proliposomes[5]

DC-Cholesterol HCl may be used in the preparation of colloidal dispersions for transmission electron microscopy studies.[1] It may also be used in liposome preparation for assessing hexokinase activity,[6] poly (lactic-coglycolicacid) loading[7] and in mRNA transfection studies.[8]

Biochem/physiol Actions

DC-Cholesterol is an effective transfection agent and a liposome reagent.[2] Liposome of DC-Cholesterol with dioleoylphosphatidylethanolamine (DOPE) are efficient for DNA transfection.[9] It imparts positive surface charge to the microbubbles for ultra sound based transfection of clustered regularly interspaced short palindromic repeats (CRISPRs).[3]
DC-cholesterol is a cationic lipid, known to be the most efficient in gene transfection study.[10][11]

Packaging

20 mL Clear Glass Screw Cap Vial (700001P-1g)
20 mL Clear Glass Screw Cap Vial (700001P-500mg)
5 mL Amber Glass Screw Cap Vial (700001P-200mg)
5 mL Amber Glass Screw Cap Vial (700001P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel cholesterol-based cationic lipids as transfecting agents of DNA for efficient gene delivery
Ju J, et al.
International Journal of Molecular Sciences, 16(3), 5666-5681 (2015)
Ultrasound microbubble-mediated CRISPR/Cas9 knockout of C-erbB-2 in HEC-1A cells
Cai J, et al.
The Journal of International Medical Research, 47(5), 2199-2206 (2019)
PLGA/liposome hybrid nanoparticles for short-chain ceramide delivery
Zou P, et al.
Pharmaceutical Research, 31(3), 684-693 (2014)
Design of novel proliposome formulation for antioxidant peptide, glutathione with enhanced oral bioavailability and stability
Byeon JC, et al.
Drug delivery, 26(1), 216?225-216?225 (2019)
Cationic nanoliposomes meet mRNA: efficient delivery of modified mRNA using hemocompatible and stable vectors for therapeutic applications
Michel T, et al.
Molecular Therapy, 8, 459-468 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service