Skip to Content
Merck
All Photos(1)

Documents

18-1740

Sigma-Aldrich

Methylene Blue solution alkaline according to Loeffler

suitable for bacterial staining

Synonym(s):

Methylene Blue solution, Ehrlich’s reagent III, Loeffler’s Methylene Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85
Beilstein:
3641570
MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:

availability

available only in Japan

suitability

suitable for bacterial staining

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;/h5-10H,1-4H3;1H/q+1;/p-1

InChI key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonel Steffen et al.
The British journal of ophthalmology, 98(1), 13-15 (2013-10-26)
To evaluate the diagnostic accuracy of methylene blue used as a non-invasive in vivo stain to detect ocular surface squamous neoplasia (OSSN). A test validation study was performed according to Standards for the reporting of diagnostic accuracy studies (STARD) guidelines
Nicolaas Lucius Gerlach et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 72(4), 666-671 (2014-02-01)
To establish the accuracy of cone-beam computed tomographic (CBCT) views in determining the position and diameter of the mandibular canal. Two freshly frozen cadaver heads, 1 dentate and 1 edentate, were used to acquire CBCT scans. Measurements on cross-sectional CBCT
Q R J G Tummers et al.
European journal of surgical oncology : the journal of the European Society of Surgical Oncology and the British Association of Surgical Oncology, 40(7), 850-858 (2014-05-28)
Despite recent developments in preoperative breast cancer imaging, intraoperative localization of tumor tissue can be challenging, resulting in tumor-positive resection margins during breast conserving surgery. Based on certain physicochemical similarities between Technetium((99m)Tc)-sestamibi (MIBI), an SPECT radiodiagnostic with a sensitivity of
J How et al.
Gynecologic oncology, 137(3), 436-442 (2015-04-15)
With the debate over extent of lymphadenectomy in endometrial cancer, sentinel lymph node (SLN) mapping may provide a focused approach to evaluate the most relevant lymph nodes (LN) while minimizing the complications. We evaluated SLN mapping using filtered technetium(99), indocyanine
Bram Vivijs et al.
Applied and environmental microbiology, 80(19), 6054-6061 (2014-07-27)
Some members of the family Enterobacteriaceae ferment sugars via the mixed-acid fermentation pathway. This yields large amounts of acids, causing strong and sometimes even lethal acidification of the environment. Other family members employ the 2,3-butanediol fermentation pathway, which generates comparatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service