Skip to Content
Merck
All Photos(1)

Key Documents

28514

Sigma-Aldrich

Methylene Blue hydrate

≥95% (calc. to the dried substance)

Synonym(s):

Basic Blue 9 monohydrate ; Methylene Blue (hydrate)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$236.00
1 KG
$1,610.00

$236.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$236.00
1 KG
$1,610.00

About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S · xH2O
CAS Number:
Molecular Weight:
319.85 (anhydrous basis)
Beilstein:
3599847
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

$236.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (calc. to the dried substance)

form

powder

impurities

≤1% insoluble in ethanol

loss

≤22% loss on drying, 105 °C

color

very dark green

solubility

ethanol: insoluble ≤1%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O.[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH.H2O/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;;/h5-10H,1-4H3;1H;1H2/q+1;;/p-1

InChI key

WQVSELLRAGBDLX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Methylene blue is bis(dimethylamino)phenothiazine hydrochloride (tetramethylthionine),
C16H18N3SCl. It is a major basic histological dye in pharmacology and microbiology. This heterocyclic aromatic chemical appears as a dark green solid powder, changing to blue in an aqueous solution. Methylene blue is tested for histological and and bacteriological staining, and as a constituent of Wright’s blood stain.

Application

Methylene blue is widely used in several routine staining applications including Mallory’s phloxine-methylene blue on paraffin sections of tissue fixed in Zenker’s fluid for nuclear staining and bacterial staining in milk. It is also suitable for manufacturing the polychrome methylene blue used in the preparation of Romanowsky stains.Methylene Blue hydrate has also been used:
  • in the preparation of methylene blue (MB) solution
  • to stain zymograms -to label the injection site in the tissue slices
  • to pre-treat mice via oral gavage to study its effects on noise-induced hearing loss

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Guo et al.
Acta pharmacologica Sinica, 41(3), 423-431 (2019-06-15)
Indoleamine 2,3-dioxygenase 1 (IDO1) is emerging as a promising therapeutic target for the treatment of malignant tumors characterized by dysregulated tryptophan metabolism. However, the antitumor efficacy of existing small-molecule IDO1 inhibitors is still unsatisfactory, and the underlying mechanism remains largely
Paul J Derry et al.
Nanoscale, 11(22), 10791-10807 (2019-05-28)
Previously, our group reported on the promising efficacy of poly(ethylene glycol)-hydrophilic carbon clusters (PEG-HCCs) to work as broadly active and high capacity antioxidants in brain ischemia and injury models including stroke and traumatic brain injury coupled with hemorrhagic shock. PEG-HCCs

Questions

  1. Are any of the methylene blue products considered API or suitable for use in manufacturing medical devices? The products offered for sale are for Research Use Only or for In Vitro Diagnostic Testing? Additionally, what are the differences between methylene blue chloride and methylene blue zinc chloride double salt offered?

    1 answer
    1. None of the methylene blue products are considered API or suitable for use in manufacturing medical devices. They are offered for Research Use Only or for In Vitro Diagnostic Testing. Methylene blue is offered as methylene blue chloride and as a methylene blue zinc chloride double salt, each with slightly different formulas and molecular weights. Methylene blue chloride is the form of the dye used in most biological staining applications and medicinal uses. It is recommended to source methylene blue from approved pharmaceutical suppliers, especially considering the availability of methylene blue USP grade.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service