Merck
  • Identification of ocular surface squamous neoplasia by in vivo staining with methylene blue.
Identification of ocular surface squamous neoplasia by in vivo staining with methylene blue.
The British journal of ophthalmology (2013-10-26)
Jonel Steffen, James Rice, Karin Lecuona, Henri Carrara
ABSTRACT
To evaluate the diagnostic accuracy of methylene blue used as a non-invasive in vivo stain to detect ocular surface squamous neoplasia (OSSN). A test validation study was performed according to Standards for the reporting of diagnostic accuracy studies (STARD) guidelines on 75 consecutive patients who presented with ocular surface lesions suspicious of OSSN. Methylene blue 1% was instilled in vivo following local anaesthetic. Stain results were documented photographically and read by an independent observer. Lesions were excised at the same visit and evaluated histologically by pathologists who were blind to the stain results. Sensitivity, specificity, positive and negative predictive values were determined. Thirty-three patients had histologically malignant lesions, of which 32 stained with methylene blue, and 42 patients had benign or premalignant lesions, of which 21 stained with methylene blue. Methylene blue had a sensitivity of 97%, specificity of 50% and positive and negative predictive values of 60% and 96%, respectively. The topical application of methylene blue is a simple, inexpensive, non-invasive diagnostic test that can be helpful in excluding malignant ocular surface lesions but cannot replace histology as gold standard for diagnosis of OSSN.
Sigma-Aldrich
Methylene Blue solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution