Skip to Content
Merck
All Photos(1)

Key Documents

11412272001

Roche

Transforming Growth Factor-β1, human (hTGF-β1)

recombinant (CHO cells)

Synonym(s):

hTGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in CHO cells

Assay

>95% (SDS-PAGE)

form

solution

potency

<0.1 ng/mL EC50

mol wt

25,000 Da

packaging

pkg of 1 mL (1 μg)

manufacturer/tradename

Roche

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.1 EU/μgtested (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TGFB1(7040)

General description

Recombinant, human TGF- β1 is produced in CHO (Chinese Hamster Ovary) cells and purified by standard chromatographic techniques.
Solution in PBS and 1 mg/ml BSA, filtered through 0.2 μm pore size membrane.

Specificity

Species specificity: Transforming growth factor-β1 (TGF-β1) is effective on human and mouse cells.
Specific activity/EC50: <0.1 ng/ml, at least the same specific activity (EC<SUB>50) compared to the indicated standard is guaranteed.

Application

Transforming growth factor-β1, human (hTGF-β1) has been used to explore function and underlying mechanism of periostin (PN) during orthodontic periodontal ligament remodeling.[1]

Biochem/physiol Actions

Transforming Growth Factor-β1, human (hTGF-β1), is a multifunctional growth factor affecting many functions in a variety of different cells.[2]
Transforming growth factor-β (TGF-β) family comprises of a group of multifunctional cytokines which are involved in controlling apoptosis, growth and differentiation of cells, and play important role during embryo development.[3]

Quality

Purity: >95% (SDS-PAGE)
Endotoxin level: <0.1 EU/μg (LAL-test), <10 EU/ml (LAL-test)
(Note: 1 EU corresponds to 0.1 ng)

Sequence

Chain Length 2 x 112 AA
Two identical polypeptide chains (112 amino acids) connected by disulfide bridges, identical to natural hTGF-β1.

Unit Definition

EC50 definition: The concentration of hTGF-β1 that is required to mediate half maximal inhibition of DNA synthesis (BrdU incorporation) with Mv1-Lu cells.

Preparation Note

Working concentration: 0.01 - 10 ng/ml
Recombinant, human TGF-β 1 exerts its biological activity in the concentration range of 0.01–10 ng/ml.
Working solution: Dilute the concentrated TGF-β1 solution (1 μg/ml) with PBS or culture medium containing BSA [or HSA (human serum albumin)], 1 mg/ml (0.1%) or serum 1–10%.
Storage conditions (working solution): For best results, prepare appropriate aliquots and store at -15 to -25 °C.
Note: Avoid repeated freezing and thawing.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

α-MSH stimulation contributes to TGF-β1 production via MC1R-MITF signaling pathway in melanoma cell
Hayashi E, et al.
Inflammation and regeneration, 35(5), 244-254 (2015)
Periostin is essential for periodontal ligament remodeling during orthodontic treatment.
Xu HY, et al.
Molecular Medicine Reports, 15(4), 1800-1806 (2017)
Cloning, expression, and characterization of chicken transforming growth factor ?4
Pan, Hongjie, and Jaroslava Halper
Biochemical and Biophysical Research Communications, 24-30 (2003)
Barbara Chiavarina et al.
Theranostics, 11(4), 1626-1640 (2021-01-08)
Colorectal cancer (CRC) cells are traditionally considered unresponsive to TGFβ due to mutations in the receptors and/or downstream signaling molecules. TGFβ influences CRC cells only indirectly via stromal cells, such as cancer-associated fibroblasts. However, CRC cell ability to directly respond

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service