Skip to Content
Merck
All Photos(1)

Key Documents

GF346

Sigma-Aldrich

Transforming Growth Factor-β1 human

Animal free, >98%, recombinant, expressed in CHO cells, suitable for cell culture

Synonym(s):

TGFb1, TGFbI, TGFB, Transforming Growth Factor-β1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

Product Name

TGF-β1,
Human Recombinant Animal Free,

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

Product Source: CHO cells
The three mammalian isoforms of TGF-β (TGF-β1, TGF-β2, TGF-β3) signal through the same receptor and elicit similar biological responses. They are multifunctional cytokines that regulate cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix. They are involved in various physiological processes including embryogenesis, tissue remodeling and wound healing. They are secreted predominantly as latent complexes which are stored at the cell surface and in the extracellular matrix. The release of biologically active TGF- isoform from a latent complex involves proteolytic processing of the complex and /or induction of conformational changes by proteins such as thrombospondin-1. TGF-β1 is the most abundant isoform secreted by almost every cell type. It was originally identified for its ability to induce phenotypic transformation of fibroblasts and recently it has been implicated in the formation of skin tumors.

Recombinant human TGF-β1 is a 25.0 kDa protein composed of two identical 112 amino acid polypeptide chains linked by a single disulfide bond.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Determined by TGF-β1’s ability to inhibit the mouse IL-4-dependent proliferation of mouse HT-2 cells. The expected ED50 is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 10^7 units/mg.

Physical form

Product is presented without additives and is filtered through a 0.2 micron filter before lyophilization

Storage and Stability

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avijit Dutta et al.
Communications biology, 6(1), 600-600 (2023-06-04)
Excessive inflammation is a postulated cause of severe disease and death in respiratory virus infections. In response to severe influenza virus infection, adoptively transferred naïve hemagglutinin-specific CD4+ T cells from CD4+ TCR-transgenic 6.5 mice drive an IFN-γ-producing Th1 response in
Weili Ge et al.
Molecular medicine reports, 21(1), 478-484 (2020-01-16)
Endothelial to mesenchymal transition (EndMT) has been confirmed to participate in several cardiovascular diseases. In addition, EndMT of circulating endothelial cells (CECs) contributes to the pathology of musculoskeletal injury. However, little is known about the molecular mechanism of CECs undergoing
Ludong Zhang et al.
Artificial cells, nanomedicine, and biotechnology, 48(1), 648-655 (2020-02-20)
Recently, the functions of circular RNAs (circRNAs) on cancer initiation and development arouse wide concern. Herein, we tested the influences of circ-ZNF652 on renal carcinoma cell growth and metastasis. Firstly, clinical renal carcinoma tissues and corresponding normal tissues were collected.
Guohui Gao et al.
Biology, 10(7) (2021-07-03)
TGF-β signaling plays a pivotal role in promoting tumor cell migration and cancer metastasis. ΔNp63α and TAp63α are two major isoforms of p53-related p63 protein. Our recent study has shown that TGF-β1 promotes ΔNp63α protein degradation to facilitate cancer metastasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service