Skip to Content
Merck
All Photos(1)

Key Documents

TX0282

Supelco

Tetrahydrofuran

≥99.9% (GC), contains 0.0140--0.0310% BHT as stabilizer, non-UV, suitable for LC/MS, gas chromatography (GC) and HPLC, OmniSolv®

Synonym(s):

Tetrahydrofuran, Diethylene oxide, THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-726-8
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Tetrahydrofuran Non-UV, OmniSolv®

vapor density

2.5 (vs air)

Quality Level

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

610 °F

contains

0.0140--0.0310% BHT as stabilizer

expl. lim.

1.8-11.8 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.01% Peroxide (as H2O2)
≤0.03% Water (H2O)
≤1.0 μeq/g Titrable acid

color

APHA: ≤10

refractive index

n20/D 1.407 (lit.)

pH

7-8 (20 °C, 200 g/L in H2O)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

transition temp

flash point -21.5 °C

density

0.889 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniSolv®
Stabilized with 250 ppm BHT. Not Optically Transparent in UV

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minColor (APHA)10 maxFiltered through 0.2 µm filterTo pass testFormClear liquidIdentity (IR-spectrum)ConformsPeroxide (as H2O2)0.01% maxRefractive index (at 20°C)Stabilizer (BHT)0.0140-0.0310%Titrable acid1.0µeq/g maxWater (H2O)0.03% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service