Key Documents
Z290459
Sigma-Aldrich
Vacuum filtration assembly flasks
side-arm flask, capacity 1000 mL
Synonym(s):
Filtration assembly
Select a Size
1 EA
$141.00
$141.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$141.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.24
$141.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
sterility
non-sterile
packaging
pack of 1
pkg of 1 ea
manufacturer/tradename
Sigma-Aldrich
capacity
1000 mL
fitting
connector for stopper
Choose from one of the most recent versions:
Certificates of Analysis (COA)