GG-1L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities
BIOSCOT™ Anti-D from the clone RUM-1, Human IgM, Blood Grouping Reagent
Synonym(s):
Anti-D (clone RUM-1) Blood Grouping Reagent
About This Item
Recommended Products
biological source
human
Quality Level
antibody form
unpurified
clone
RUM-1, monoclonal
form
liquid
packaging
pkg of 1 L
manufacturer/tradename
BIOSCOT™
concentration
25-40 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.6-7.0
isotype
IgM
shipped in
ambient
storage temp.
2-8°C
General description
Application
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service