Key Documents
Z516856
Sigma-Aldrich
Aldrich® two-neck round-bottom flask
capacity 100 mL, Joints: ST/NS 24/40 (2)
Synonym(s):
bottom flask, flask round, round flask
Select a Size
1 EA
$141.00
$141.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$141.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$141.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
round bottom flask
two-neck
Joint
Joints: ST/NS24/40 (2)
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
100 mL
General description
Angled ST joints. Z516929 and Z516953 have 24/40 side joints.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)