Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB193F

Sigma-Aldrich

Milli-Mark® Anti-CD33-FITC Antibody, clone WM-54

clone WM-54, Milli-Mark®, from mouse

Synonym(s):

Myeloid cell surface antigen CD33, Sialic acid-binding Ig-like lectin 3, Siglec-3, gp67

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

WM-54, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD33(945)

General description

CD33 is a member of the siglec family (sialic acid binding immunoglobulin-like lectins) referred to as Siglec-3 (Freeman SD, Kelm S, Barber EK and Crocker PR, 1995, Blood, 2005-2012). It is the smallest member of the siglec family having a molecular weight of 67 kDa (Peiper SC, Ashmun RA and Look TA (1988) Blood., 72: 314-321). CD33 mediates protein-carbohydrate cellular interactions in addition to protein-protein recognition (Crocker, 2002). The CD33 gene is located on chromosome 19 (Peiper, 1997). CD33 is expressed by subsets of monocytes and neutrophils. CD33 is the major antigen to differentially diagnose myeloid from lymphoid leukemias, as it is expressed on 90% of myeloid and not lymphoid leukemic cells (Andrews 1989; Griffin 1984; and Sabbath 1986). Committed hemapoietic progenitors, including multipotential stem cells (CFU-GEMM) and precursors of granulocytes and macrophages (CFU-GM) and early erythroid progenitors (BFU-E) express CD33 (Peiper, 1989 and 1997). Anti-CD33, WM-54, was included in the Fourth International Workshop and Conference on Human Leucocyte Differentiation Antigens and studies by a number of laboratories confirmed its reactivity with CD33 (Peiper, 1989).

Specificity

Antibody recognizes Human CD33 antigen.

Immunogen

Leukemic cells from a patient with AML.

Application

Milli-Mark Anti-CD33-FITC Antibody, clone WM-54 detects level of CD33 & has been published & validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human PBMCs

Target description

67 kDa (Peiper, 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
PBMCs

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service