Skip to Content
Merck
All Photos(1)

Key Documents

15862

Sigma-Aldrich

Cobalt chloride 0.1 M solution

Synonym(s):

Cobalt(II)-chloride solution, Additive Screening Solution 24/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.32
grade:
for analytical purposes
Pricing and availability is not currently available.

grade

for analytical purposes

density

1.01 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cl[Co]Cl

InChI

1S/2ClH.Co/h2*1H;/q;;+2/p-2

InChI key

GVPFVAHMJGGAJG-UHFFFAOYSA-L

Application

Cobalt chloride 0.1M solution has been used to simulate hypoxic exposure and HIF-1 (hypoxia-inducible factor 1) expression in prostate cancer cells.[1] It has been used to induce hypoxia in human intestinal epithelial cells.[2]

Biochem/physiol Actions

Cobalt chloride induces hypoxia condition in cells by upregulating hypoxia-inducible factor-1α (HIF-1α), erythropoietin and glycolytic enzymes. It is also responsible for mitochondrial DNA damage in rat neuronal PC12 cells.[3] It is also responsible for the induction of apoptosis.[4]
Cobalt chloride 0.1M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulation of duodenal SLC transporters and activation of proinflammatory signaling constitute the early response to high altitude in humans.
Wojtal KA, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 307, G673-G673 (2014)
Francesca Lazzara et al.
Frontiers in pharmacology, 11, 1063-1063 (2020-08-28)
Retinal hypoxia is one of the causative factors of diabetic retinopathy and is also one of the triggers of VEGF release. We hypothesized that specific dysregulated miRNAs in diabetic retinopathy could be linked to hypoxia-induced damage in human retinal endothelial
Exposure to hypoxia following irradiation increases radioresistance in prostate cancer cells.
Hennessey D, et al.
Urologic Oncology, 31, 1106-1106 (2013)
Mitochondrial DNA damage and a hypoxic response are induced by CoCl(2) in rat neuronal PC12 cells.
Wang G, et al.
Nucleic Acids Research, 28, 2135-2135 (2000)
CoCl(2) induces apoptosis via the 18 kDa translocator protein in U118MG human glioblastoma cells.
Zeno S, et al.
Biochemistry, 48, 4652-4652 (2009)

Questions

  1. How long can this be stored for at 2-8 degrees?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service