Skip to Content
Merck
All Photos(3)

Key Documents

340855

Sigma-Aldrich

Glutaraldehyde solution

50 wt. % in H2O

Synonym(s):

Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.21
form:
liquid

vapor density

1.05 (vs air)

Quality Level

vapor pressure

15 mmHg ( 20 °C)

form

liquid

concentration

50 wt. % in H2O

technique(s)

electron microscopy: suitable
immunoblotting: suitable
immunohistochemistry: suitable

color

colorless

refractive index

n20/D 1.42

mp

-21  °C ((-6 °F))

solubility

water: soluble

density

1.106 g/mL at 25 °C

suitability

suitable for microfluidics/nanofluidics

application(s)

cell analysis
genomic analysis
life science and biopharma
sample preparation

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutaraldehyde (Glutaric dialdehyde, Pentane-1,5-dial) is an aliphatic dialdehyde that can be employed as an effective biochemical reagent for various purposes such as a protein crosslinker, enzyme immobilization in microscopy, histochemistry, and cytochemistry. It serves as an amine-reactive homobifunctional crosslinker and a cell fixative before SDS-PAGE, staining, or electron microscopy, making it suitable for a wide range of morphological studies.

Glutaraldehyde is capable of efficiently crosslinking amine and hydrazine derivatives to proteins and other amine-containing polymers. Notably, in certain reactions, biotin hydrazides have been directly coupled to nucleic acids using glutaraldehyde, presenting a potential utility in conjugating fluorescent hydrazides and hydroxylamines to DNA. In histology studies, Glutaraldehyde is also employed to preserve tissue sections and prepare them for closer examination.

Application

Cross-linking agent for gelatin, poly(vinyl alcohol), and polyheptapeptides.
Glutaraldehyde may be used in the following studies:
  • To compose the fixative solution (Glutaraldehyde + Paraformaldehyde + NaPO4) for use in high-resolution light microscopy and electron microscopy studies.
  • To study the conjugation of goat anti-horseradish peroxidase with alkaline phosphatase by a reported method.
  • To compose the primary fixative, which is employed to protect the deterioration of cytoplasmic features of yeast cells during permanganate fixation.

Features and Benefits

  • Versatile and adaptable for wide variety of research applications
  • Ready-made solution reduces the need for preparation time

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Biomimetics, 1, 219-219 (1992)
Chem. Abstr., 121, 52250t-52250t (1994)
Polymer Journal (1993)
Minutes Int. Symp. Microencapsulation, 9th, 191-191 (1993)
Preservation of fine structures in yeast by fixation in a dimethyl sulfoxide-acrolein-glutaraldehyde solution.
D W Schwab et al.
Stain technology, 45(4), 143-147 (1970-07-01)

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service