Skip to Content
Merck
All Photos(1)

Key Documents

CC123

Sigma-Aldrich

Caspase 8, recombinant human active

Synonym(s):

FLICE, MASH, Mch5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

recombinant

expressed in E. coli

form

solid

specific activity

25 units

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

General description

Caspase-8 is a member of the interleukin-1β converting enzyme (ICE) family of cysteine proteases. As with other caspases, caspase-8 also exists in cells as an inactive proenzyme. During apoptosis procaspase-8 is processed at aspartate residues by self-proteolysis and/or cleavage by another caspase. The processed form of caspase-8 consists of large and small subunits which associate to form the active enzyme. The active caspase-8 has been shown to be involved in the proteolysis of poly (ADP-ribose) polymerase (PARP), an enzyme that is involved in DNA repair and genomic maintenance.
The active recombinant human caspase-8 was expressed in E. coli. The expressed caspase-8 spontaneously undergoes autoprocessing to yield the subunits characteristic of the active enzyme. The recombinant caspase-8 preferentially cleaves the substrates consisting of consensus sequence IETD (e.g., IETD-AFC and IETD-pNA). The recombinant enzyme can also be used as a positive control in caspase assays or in determining the specificty of caspase substrates.Recommended usage: 0.5-1 unit for fluorometric caspase assays and 2-4 units for colorimetric caspase assays. Optimal working dilutions must be determined by end user.

Application

Recommended usage: 0.5-1 unit for fluorometric caspase assays and 2-4 units for colorimetric caspase assays.

Optimal working dilutions must be determined by end user.

Unit Definition

Specific Activity: Where one unit of the recombinant caspase-8 is the enzyme activity that cleaves 1 nmol of the caspase substrate IETD-pNA (pNA: pnitroanaline) per hour at 37°C in a reaction solution containing 50 mM HEPES, 50 mM NaCl, 0.1% CHAPS, 10 mM EDTA, 5% Glycerol, and 10 mM DTT.

Physical form

Lyophilized. Reconstitute with 25 μl PBS containing 15% glycerol.

Storage and Stability

Maintain at -70°C for 3-6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specific caspase pathways are activated in the two stages of cerebral infarction.
A Benchoua,C Guegan,C Couriaud,H Hosseini,N Sampaio,D Morin,B Onteniente
The Journal of Neuroscience null
Jiro Fujita et al.
Microbiology and immunology, 47(6), 447-451 (2003-08-09)
In a previous study, we demonstrated anti-vimentin antibodies in sera of patients with interstitial pneumonia. We hypothesized that antibodies in sera might detect vimentin fragments formed during the process of apoptosis. To prove this, recombinant human vimentin was digested by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service