Skip to Content
Merck
All Photos(3)

Key Documents

SAE1000

Sigma-Aldrich

SARS-CoV-2 Receptor Binding Domain

Spike protein RBD recombinant, expressed in HEK 293 cells

Synonym(s):

COVID-19, SARS-CoV-2, Spike Protein RBD, Spike-ACE2, coronavirus

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$503.00

$503.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$503.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$503.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

~35 kDa

technique(s)

ELISA: 2 μg/mL (({Functional} RBD binds 50% of the optimal binding RBD-ACE2 ))

shipped in

ambient

storage temp.

−20°C

Related Categories

General description

SARS-CoV-2 belongs to Coronaviridae (CoVs) - a large family of viruses that usually cause mild to moderate upper-respiratory tract diseases.1 CoVs are single-stranded RNA viruses. They have a high mutation rate and are therefore highly diverse.1 Their hosts include human and non-human mammals, as well as birds.1

Specificity

This product is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of 25 kDa (amino acids 319- 541) and includes a C-terminal polyhistidine tag.

Application

The activity of this product is tested by a functional ELISA. Immobilization of Spike protein Receptor Binding Domain at 2 μg/mL (100 μL/well), allows binding of ACE2, biotin tagged (Cat. No. SAE0171). The half maximal binding (binding which produces 50% of the optimal binding response) is ≤2 μg/mL.

Biochem/physiol Actions

SARS-CoV-2 infects cells which express angiotensin converting enzyme 2 (ACE2).2 ACE2 binds to the viral envelope-associated spike (S) protein as a cellular receptor, following proteolytic cleavage of both S and ACE2 by serine proteases.3 The receptor binding domain (RBD) of S protein is the domain that specifically binds to ACE2.

Physical form

This product is supplied as a lyophilized powder in phosphate buffered saline (PBS), pH 7.4, with no carrier proteins.

Storage and Stability

Store the product at –20 °C. The product retains its activity for at least 2 years as supplied. After initial thawing, it is recommended to store the protein in working aliquots at –20 °C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Data presented is the available current product information and provided as-is. This product has not been tested or verified in any additional applications, sample types, including any clinical use. Experimental conditions must be empirically derived by the user. Our Antibody Guarantee only covers tested applications stated herein and conditions presented in our product information and is not extended to publications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, I cannot find the user guidelines for this product. It will be helpful if anyone can describe the application of this product in detail. It's a little bit confusing for me. Thank you in advance.

    1 answer
    1. The Datasheet for this product can be accessed in the DOCUMENTATION section under the 'More Documents' tab. Please see the product Datasheet below:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/291/243/sae1000dat-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service