Skip to Content
Merck
All Photos(1)

Key Documents

AP307F

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, (H+L) FITC conjugate

1.0 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

1.0 mg/mL

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

The reagent is an affinity purified antibody from goat. The purified antibody is conjugated to fluorescein isothiocyanate (FITC) and stabilized in buffer

Application

Goat anti-Rabbit IgG Antibody, (H+L) FITC conjugate detects level of Rabbit IgG & has been published & validated for use in IF.
Immunohistochemistry:1:200-1:500 (1-4)

Immunocytochemistry: 1:200-1:500 (1,4)

Flow Cytometry: 1 μg per 1x10E6 cells (1,2,4)

Optimal working dilutions must be determined by the end user.

Storage and Stability

The undiluted antibody is stable at 2–8°C for 12 months. Do not store in a diluted format.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuichi Shibuya et al.
Nutrients, 12(2) (2020-02-26)
Acai (Euterpe oleracea Mart. Palmae, Arecaceae) is a palm plant native to the Brazilian Amazon. It contains many nutrients, such as polyphenols, iron, vitamin E, and unsaturated fatty acids, so in recent years, many of the antioxidant and anti-inflammatory effects
Raheleh Hatami et al.
Science translational medicine, 5(169), 169ra12-169ra12 (2013-01-25)
Metastasis is the major cause of cancer mortality. A more thorough understanding of the mechanisms driving this complex multistep process will aid in the identification and characterization of therapeutically targetable genetic drivers of disease progression. We demonstrate that KLF6-SV1, an
Bin Huang et al.
Technology in cancer research & treatment, 18, 1533033819879905-1533033819879905 (2019-11-24)
Hepatocellular carcinoma is a highly vascularized tumor, so it is critical to study its angiogenesis. Cancer-associated fibroblasts and enhancer of zeste homolog 2 play an important role in tumor angiogenesis and became significant hallmarks of cancer. But the relationship between
Dong Zhou et al.
Veterinary research, 46, 95-95 (2015-09-19)
Although breeding of F4 receptor - negative (F4R(-)) pigs may prevent post-weaning diarrhea, the underlying immunity is poorly understood. Here, various doses of a Bacillus licheniformis and Bacillus subtilis mixture (BLS-mix) were orally administered to F4ab/acR(-) pigs for 1 week before F4 (K88) - positive
Jinbin Zhai et al.
eLife, 10 (2021-05-05)
A major barrier to intraspinal regeneration after dorsal root (DR) injury is the DR entry zone (DREZ), the CNS/PNS interface. DR axons stop regenerating at the DREZ, even if regenerative capacity is increased by a nerve conditioning lesion. This potent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service