Skip to Content
Merck
All Photos(1)

Key Documents

AP124F

Sigma-Aldrich

Goat Anti-Mouse IgG Antibody, (H+L) FITC Conjugated

2 mg/mL (after reconstitution), Chemicon®

Synonym(s):

Goat Anti-Mouse IgG FITC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

2 mg/mL (after reconstitution)

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Immunoglobulin G (IgG) is a multimeric immunoglobulin, built of two heavy chains γ and two light chains. Each complex has two antigen binding sites. It is one of the most abundant proteins in human serum with normal levels between 8-17 mg/ml in adult blood. IgG is important for our defense against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. Furthermore, this is the only isotype that can pass through the human placenta, thereby providing protection to the fetus in its first weeks of life before its own immune system has developed. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Affinity purified antibody shows a single band when run in an IEP system at a minimum concentration of 10 mg/mL using whole mouse serum as the antigen.
Purity:
Affinity purified antibody shows a single band against Rabbit anti Goat Serum when run as the antigen in IEP at a minimum concentration of 10 mg/mL.

Application

Affinity purified antibody shows a single band against Rabbit anti-Goat Serum when run as the antigen in IEP at a minimum concentration of 10 mg/mL.

Suggested dilution for most applications: 1:50-1:200.

Optimal working dilutions must be determined by the end user.
Goat anti-Mouse IgG Antibody, (H+L) FITC Conjugated is an antibody against Mouse IgG for use in IF.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Affinity purified antibody shows a single band against Rabbit anti Goat Serum when run as the antigen in IEP at a minimum concentration of 10 mg/mL.

Physical form

ImmunoAffinity Purified
Lyophilized from 0.02 M phosphate buffered saline, 0.25 M sodium chloride, pH 7.6, with 0.1% sodium azide and 15 mg/mL bovine serum albumin.
F/P Ratio:
14.5 μg FITC per mg of antibody

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months after date of receipt. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles. Protect from light.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Howe et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 2(5), 485-495 (2000-11-18)
The oncolytic effect of adenoviruses may provide an efficient means to destroy tumor tissue if viruses could be developed with sufficient selectivity and efficacy. In this report we have characterized several adenoviruses, each with different mutations in the E1 region
Efficacy of optimized in vitro predegeneration period on the cell count and purity of canine Schwann cell cultures.
Niapour, N; Mohammadi-Ghalehbin, B; Golmohammadi, MG; Amani, M; Salehi, H; Niapour, A
Iranian Journal of Basic Medical Sciences null
Reduction of Aedes aegypti vector competence for dengue virus under large temperature fluctuations.
Carrington, LB; Seifert, SN; Armijos, MV; Lambrechts, L; Scott, TW
The American Journal of Tropical Medicine and Hygiene null
Farnoosh Jafarpour et al.
International journal of fertility & sterility, 4(4), 148-155 (2011-01-01)
Reconstructed embryos from terminally differentiated somatic cells have revealed high levels of genomic methylation which results in inappropriate expression patterns of imprinted and non-imprinted genes. These aberrant expressions are probably responsible for different abnormalities during the development of clones. Improvement
Pao-Pao Yang et al.
Journal of biomedical science, 22, 81-81 (2015-09-24)
Neuropathic pain is a very troublesome and difficult pain to treat. Although opioids are the best analgesics for cancer and surgical pain in clinic, only oxycodone among opioids shows better efficacy to alleviate neuropathic pain. However, many side effects associated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service