8.00605
Propionic acid
for synthesis
Synonym(s):
Propionic acid, Methyl acetic acid
About This Item
Recommended Products
vapor pressure
5 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
form
liquid
autoignition temp.
485 °C (DIN 51794)
potency
2600 mg/kg LD50, oral (Rat)
500 mg/kg LD50, skin (Rabbit)
expl. lim.
2.1-12 % (v/v)
pH
2.5 (20 °C, 100 g/L in H2O)
bp
140.7-141.4 °C/1013 hPa
mp
-20 °C
transition temp
flash point 50.5 °C
density
0.993 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
OC(=O)CC
InChI
1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)
InChI key
XBDQKXXYIPTUBI-UHFFFAOYSA-N
Related Categories
Analysis Note
Density (d 20 °C/ 4 °C): 0.993 - 0.994
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
129.2 °F - closed cup
Flash Point(C)
54 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service