Skip to Content
Merck
All Photos(2)

Key Documents

N10950

Sigma-Aldrich

Nitrobenzene

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
507540
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
210-211 °C (lit.)
vapor pressure:
0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

4.2 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

899 °F

expl. lim.

40 %

dilution

(for general lab use)

refractive index

n20/D 1.551 (lit.)

pH

8-8.5 (20 °C, 1 g/L)

bp

210-211 °C (lit.)

mp

5-6 °C (lit.)

density

1.196 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

LQNUZADURLCDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitrobenzene is an aromatic compound that can be prepared by the nitration of benzene with nitric acid and sulfuric acid mixture.[1] Reduced graphene oxide catalyzed reduction of nitrobenzene has been reported to form N-phenylhydroxylamine as an intermediate.[2] The ability of nitrobenzene to cause DNA damage has been assessed in tobacco by single-cell gel electrophoresis (SCGE). The mechanism of photolysis of nitrobenzene has been reported.[3]

Application

Nitrobenzene may be used as an oxidizing agent in the oxidative carbonylation of methanol in the presence of palladium diphosphane complexes.[4]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced graphene oxide as a catalyst for hydrogenation of nitrobenzene at room temperature.
Gao Y, et al.
Chemical Communications (Cambridge, England), 47(8), 2432-2434 (2011)
Photodissociation dynamics of nitrobenzene and o-nitrotoluene.
Lin MF, et al.
J. Chem. Phys. , 126(6), 064310-064310 (2007)
Nitrobenzene and Nitrotoluenes.
Dugal, M.
Kirk-Othmer Encyclopedia of Chemical Technology (2005)
Mechanistic Study of the Oxidative Carbonylation of Methanol Catalyzed by Palladium Diphosphane Complexes with Nitrobenzene as Oxidant.
Mooibroek TJ, et al.
European Journal of Inorganic Chemistry, 2012(9), 1403-1412 (2012)
Wataru Kosaka et al.
Chemical communications (Cambridge, England), 49(16), 1594-1596 (2012-11-24)
A porous one-dimensional coordination chain compound, [Ru(2)(p-F-PhCO(2))(4)(phz)] (; p-F-PhCO(2)(-) = para-fluorobenzoate; phz = phenazine) derived by drying its nitrobenzene-solvated compound, specifically adsorbs CO(2) at 195 K in a stepwise sorption manner.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service