Skip to Content
Merck
All Photos(1)

Key Documents

18104

Sigma-Aldrich

Calcium propionate

99.0-100.5% (in dried substance), meets analytical specification of E 282

Synonym(s):

Propionic acid calcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$102.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$102.00

About This Item

Linear Formula:
(CH3CH2COO)2Ca
CAS Number:
Molecular Weight:
186.22
Beilstein:
3698682
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.0-100.5% (in dried substance)

form

solid

quality

meets analytical specification of E 282

impurities

≤0.3% water insolubles
≤10 mg/kg heavy metal (as Pb)

loss

≤4% loss on drying, 105 °C, 4 h

pH

6.0-9.0 (20 °C, 10%)
7.5-9.0 (20 °C, 10%)

anion traces

fluoride (F-): ≤10 mg/kg

cation traces

As: ≤3 mg/kg
Fe: ≤50 mg/kg
Hg: ≤1 mg/kg
Pb: ≤5 mg/kg

SMILES string

CCC(=O)O[Ca]OC(=O)CC

InChI

1S/2C3H6O2.Ca/c2*1-2-3(4)5;/h2*2H2,1H3,(H,4,5);/q;;+2/p-2

InChI key

BCZXFFBUYPCTSJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serkan Yilmaz et al.
Toxicology and industrial health, 30(10), 926-937 (2012-11-29)
In vitro genotoxic effects of antioxidant additives, such as citric acid (CA) and phosphoric acid (PA) and their combination, as well as antimicrobial additives, such as benzoic acid (BA) and calcium propionate (CP), on human lymphocytes were determined using alkaline
Markus C E Belz et al.
Applied microbiology and biotechnology, 96(2), 493-501 (2012-05-10)
The consumption of low-salt bread represents an efficient way to improve public health by decreasing cardiovascular health issues related to increased intakes of sodium chloride (NaCl). The reduction of NaCl influences the bread quality characteristics, in particular the shelf-life. Calcium
Hugo Alarie et al.
International journal of pharmaceutics, 569, 118552-118552 (2019-08-04)
This project aimed to develop a suspension vehicle specifically designed for pediatric use. Excipients were selected according to their safety and recorded use in pediatrics. Two suspension vehicles were formulated at neutral and acidic pH. A few compositions were defined
N H Kim et al.
Journal of applied microbiology, 119(1), 139-148 (2015-03-31)
To develop a mild blanching method with calcium salts to ensure microbiological safety and quality of fresh-cut spinach. The antimicrobial efficacy of eight calcium salts was evaluated on Escherichia coli O157:H7 at 45-65°C and calcium hydroxide (Ca(OH)2 ) showed the
Dominic Dussault et al.
Food microbiology, 53(Pt B), 115-121 (2015-12-19)
The objective of this study was to evaluate the impact of seven independent factors consisting of sodium nitrite, pH, sodium chloride, sodium acetate, sodium lactate syrup, calcium propionate and a blend of nisin and hop alpha acids on the growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service