Skip to Content
Merck
All Photos(2)

Key Documents

79143BC

Millipore

GDS PPMX Chime Box

BioControl Systems, for use with GDS PickPen® PIPETMAX® System

Synonym(s):

Acoustic Alert System, Audible Signaling Device, Audio Alert Module, Audio Cue System, Chime-based Alert System, Pipetting Process Notifier, Pipetting Workflow Auditory Device, Pipetting Workflow Chime Box, Pipetting workflow, Sound Indicator Box, Sound Notification Box

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,380.00

$1,380.00


Estimated to ship onMay 08, 2025


Request more information

Select a Size

Change View
1 EACH
$1,380.00

About This Item

UNSPSC Code:
41121521

$1,380.00


Estimated to ship onMay 08, 2025


Request more information

packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen PIPETMAX System

storage temp.

room temp

General description

The GDS PPMX Chime Box is a component designed for use with the GDS PickPen® PIPETMAX® System. The chime box is used to provide audio notifications during protocols. It can be programmed to use any MP3 sound as the alert, helping to prevent potential mistakes and ensure data integrity.

Application

GDS PPMX Chime Box is a signaling device that alerts the user to specific steps during protocols when used with the GDS PickPen® PIPETMAX® System.

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service