Skip to Content
Merck
All Photos(5)

Key Documents

79100BC

Millipore

GDS PickPen® PIPETMAX® System

Automated reagent and sample preparation system for use with Assurance® GDS PCR system

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$61,640.00

$61,640.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
$61,640.00

About This Item

UNSPSC Code:
41121521

$61,640.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Quality Level

manufacturer/tradename

Assurance GDS® PickPen
BioControl Systems 79100

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS

storage temp.

room temp

Related Categories

General description

Assurance® GDS (Genetic Detection System) is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The GDS PickPen® PIPETMAX® (PPMX) is an automated magnetic particles and liquid handler for use with Assurance® GDS assays. The PPMX is capable of dispensing GDS reagents which include, Concentration Reagent, Wash Solution, Resuspension Buffer and select media (BHI/DFB). In addition, the PPMX has also been programmed to automate GDS PickPen® Immunomagnetic Separation (IMS) procedures and transfers to amplification tubes

Application

The GDS PickPen® PIPETMAX® (PPMX) is an automated magnetic particles and liquid handler for use with Assurance® GDS assays for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Features and Benefits

  • Flexible modular design accommodates small and large runs; maximizing productivity
  • Utilizes Air Displacement pipetting and follows an aseptic travel path minimizing cross-contamination risk
  • LIMS compatibility and barcode enabled for resource tracking
  • Instrument operated via touchscreen tablet with all Assurance GDS protocols pre-programmed
  • Step-by-step wizard guides users through run creation and sample loading
  • Small lab bench footprint - W 21.4” x D 25.8” x H 20.9” and minimal preventative maintenance
  • Supports both the GDS Rotor-Gene® standard and high throughput rotors

Components

  • PPMX Instrument with rotating cover
  • Alignment Head
  • Removable Tray 96 well
  • Pipette Tip Reload Rack
  • PickPen Tip Reload Rack
  • Tip Disposal Bin
  • Power Supply (110V/220V)
  • Power/USB Cabling kit
  • Power Cords (US and EU)
  • USB Cable
  • Back Plug, Black
  • Allen Wrench, 5 mm
  • MAX8x200 Pipette Head
  • Assurance GDS PickPen Magnetic Head
  • Assurance GDS Reagent Holder
  • Tip Mover with Tip Box Holder
  • Concentration Reagent Bottle Fill Level Sleeve
  • Chime Sound Box with hardware
  • Gel Cooling Block (2/cs)
  • Control Tablet with TRILUTION micro software pre?loaded + Charger and Tablet holder or
  • Microsoft Surface laptop with TRILUTION micro software pre-loaded + Charger and Keyboard cover/Pen

These items come with US plugs only.

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc
Rotor-Gene is a registered trademark of Qiagen GmbH

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service