Skip to Content
Merck
All Photos(2)

Key Documents

1.05459

Millipore

Tryptic Soy Broth - Dehydrated Culture Media

GranuCult® prime, USP, EP, JP, ISO 21871, granular, suitable for pathogen testing

Synonym(s):

Casein-peptone soymeal-peptone broth, Casein-peptone soyameal-peptone broth, TSB, soy broth, CASO Broth, Casein-peptone Soymeal-peptone Broth, CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

EP
ISO 21871
JP
USP

Quality Level

reg. compliance

FDA (BAM)

sterility

non-sterile

form

granular

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

solubility

30 g/L

bulk density

570 kg/m3

application(s)

microbiology
pharmaceutical

pathogen testing

storage temp.

15-25°C

suitability

aerobic bacteria (total)

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth (TSB) (also called CASO Broth or Casein-peptone Soymeal-peptone Broth) is a granulated universal culture medium, free from inhibitors and indicators, for the isolation and cultivation of fastidious aerobic bacteria, yeasts, and molds.

Application

Tryptic Soy Broth (TSB) is used for sterility testing of non-sterile preparations and products. It is used to test for specified microorganisms or total viable aerobic count determinations using the MPN method. It is also used as pre-enrichment medium for the detection of food pathogens, e.g. Salmonella, STEC.

Linkage

Replaces: 1054599027; 1.05459.5007

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanwal M Iqbal et al.
Microorganisms, 8(2) (2020-01-26)
The persistence of bacterial pathogens within environmental matrices plays an important role in the epidemiology of diseases, as well as impacts biosurveillance strategies. However, the adaptation potentials, mechanisms for survival, and ecological interactions of pathogenic bacteria such as Yersinia pestis
B Tenderis et al.
International journal of food microbiology, 321, 108560-108560 (2020-02-23)
Foodborne illnesses affect the health of consumers worldwide, and thus searching for potential antimicrobial agents against foodborne pathogens is given an increased focus. This research evaluated the influence of sodium lactate (SL), encapsulated (e) and unencapsulated (u) polyphosphates (PP; sodium

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service