Skip to Content
Merck

Skip To

73091BC

GDS PickPen®, 8-Magnets

Immunomagnetic separation device for use with Assurance® GDS PCR system

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$1,490.00

About This Item

UNSPSC Code:
41115866
NACRES:
NC.81

$1,490.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Assurance GDS®, BioControl Systems 73091

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

Assurance® GDS (Genetic Detection System) is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The proprietary GDS PickPen®, 8-Magnets is designed to transfer magnetic particles when working with Assurance® GDS pathogen detection kits. The incorporation of the PickPen® in the immunomagnetic separation (IMS) process improves the procedure by concentrating the target pathogenwhile decreasing carryover of background microflora without compromising the sensitivity.

Application

Immunomagnetic Separation (IMS)
The GDS PickPen®, 8-Magnets is designed to transfer magnetic particles when working with Assurance® GDS pathogen detection kits used for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Features and Benefits

  • Compatible with all Assurance® GDS disposable wells/plates, 8 samples can be taken through IMS at one time
  • Consistent recovery of immuno-magnetic particles during IMS
  • Lower carryover of background microflora

Other Notes

One GDS PickPen

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
79100BC73085BC73087BC
compatibility

for use with GDS Rotor-Gene

compatibility

BioControl GDS

compatibility

for use with PICKPEN®

compatibility

for use with PICKPEN®

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

manufacturer/tradename

Assurance GDS®

manufacturer/tradename

Assurance GDS® PickPen, BioControl Systems 79100

manufacturer/tradename

BioControl Systems 73085

manufacturer/tradename

BioControl Systems 73087




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters