Skip to Content
Merck
All Photos(1)

Key Documents

6073

Sigma-Aldrich

Schiff Reagent, Hotchkiss-McManus

Harleco® (For Microscopy (contains C.I. Basic Red 9))

Synonym(s):

P.A.S.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

liquid

Quality Level

manufacturer/tradename

Harleco® (For Microscopy (contains C.I. Basic Red 9))

mp

-0.1 °C

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO® For PAS staining

Application

HARLECO® For PAS staining
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
For P.A.S. Reaction. May be used as replacement for Schiff Reagent in the PAS Kit.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Met. Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service