Skip to Content
Merck
All Photos(1)

Key Documents

48900

Sigma-Aldrich

Giemsa Stain

Modified Solution

Synonym(s):

Azure eosin methylene blue, Giemsa solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$129.00
500 ML
$246.00
1 L
$457.00

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$129.00
500 ML
$246.00
1 L
$457.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
NACRES:
NA.25

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Giemsa Stain, Modified Solution, (for the staining (of cellular blood components and blood parasites))

type

(for research use only)

Quality Level

shelf life

limited shelf life

technique(s)

microbe id | staining: suitable

suitability

Chlamydia spp.
Histoplasma spp.
Plasmodium spp.
Trichomonas spp.
bacteria
protozoa
spirochetes

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tested for staining blood smears according to G. Clark (ed.), Staining Procedures, 4th edition, Williams & Wilkins, 173 (1981); together with May-Grünwald Solution for pap staining according to Pappenheim.

Caution

store at 20°C

Principle

When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration. The use of purified eosin and thiazine dyes minimizes lot-to-lot variation.

Physical form

Solution of azure B/azure II-eosin/methylene blue 1:12:2 (w/w/w) in glycerol/methanol 5:24 (v/v); total dye content: 0.6 % (w/w)

Reconstitution

Fix 10 min. in methanol; air-dry; immerse 45 min. in Giemsa Solution; rinse with distilled water; air-dry

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenő Káldy et al.
Genes, 11(7) (2020-07-10)
Two species from the families Acipenseridae and Polyodontidae, Russian sturgeon (Acipenser gueldenstaedtii, Brandt and Ratzeberg, 1833; functional tetraploid) and American paddlefish (Polyodon spathula, Walbaum 1792, functional diploid) were hybridized. The hybridization was repeated using eggs from three sturgeon and sperm
Alessandra Micera et al.
PloS one, 10(11), e0142737-e0142737 (2015-11-17)
In a previous study, we reported the upregulation of Nerve Growth Factor (NGF) and trkANGFR expression in Ocular Cicatricial Pemphigoid (OCP), an inflammatory and remodeling eye disease. Herein, we hypothesize a potential NGF-driven mechanism on fibroblasts (FBs) during OCP remodeling
Antonio Di Zazzo et al.
Journal of molecular medicine (Berlin, Germany), 98(5), 751-760 (2020-04-22)
Metabolomics has been applied to diagnose diseases, predict disease progression, and design therapeutic strategies in various areas of medicine. However, it remains to be applied to the ocular surface diseases, where biological samples are often of limited quantities. We successfully
Tobias Koeniger et al.
Stem cells (Dayton, Ohio), 39(2), 227-239 (2020-12-04)
Although the bone marrow contains most hematopoietic activity during adulthood, hematopoietic stem and progenitor cells can be recovered from various extramedullary sites. Cells with hematopoietic progenitor properties have even been reported in the adult brain under steady-state conditions, but their
Liz J Valente et al.
The Journal of cell biology, 219(11) (2020-09-05)
The mechanisms by which TP53, the most frequently mutated gene in human cancer, suppresses tumorigenesis remain unclear. p53 modulates various cellular processes, such as apoptosis and proliferation, which has led to distinct cellular mechanisms being proposed for p53-mediated tumor suppression

Questions

1–3 of 3 Questions  
  1. WITH WHICH BUFFER SHOULD I DILUTE GIEMSA SOLUTION

    1 answer
    1. Giemsa solution protocols recommend diluting with either water or a phosphate buffer. Common phosphate buffers are Weise or Sorensen's phosphate buffer. Depending upon personal preference, the pH used can vary from a low pH of 6.0 to a high of 7.2 In North America it is more common to use a Sorensen's phosphate buffer with a pH ranging from 6.8 to 7.2. A suitable catalog number for a pH 7.2 buffer is P3288. In other parts of the world slightly more acidic phosphate buffers are preferred.

      Helpful?

  2. Hi does any lab use this stain for microscopy ( haematology) ? How is it working for you on fresh peripheral blood films ?

    1 answer
    1. Giemsa solution is commonly used for hematology, histology, and parasitology labs. Peripheral blood smears and bone marrow smears are routinely stained in hematology labs. It is always preferred to use fresh peripheral blood smears or bone marrow smears when possible. The colors are more vibrant when the peripheral blood smears are stained fresh.

      Helpful?

  3. Is there a white paper available for buffer selection for Giemsa staining?

    1 answer
    1. Generally, no white papers are available. Giemsa formulations vary in pH, typically ranging between pH 6.4 to 6.9. Quality standards for stained slides differ across institutions and individuals, with varying preferences for pH buffers.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service