Skip to Content
Merck
All Photos(1)

Documents

567809-M

Millipore

Src Inhibitor, PP1

The Src Inhibitor PP1, also referenced under CAS 172889-26-8, controls the biological activity of Src. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Src Inhibitor, PP1, 4-amino-5-(4-methylphenyl)-7-(t-butyl)pyrazolo-d-3,4-pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N5
CAS Number:
Molecular Weight:
281.36
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 25 mg/mL

storage temp.

−20°C

InChI

1S/C16H19N5/c1-10-5-7-11(8-6-10)13-12-14(17)18-9-19-15(12)21(20-13)16(2,3)4/h5-9H,1-4H3,(H2,17,18,19)

InChI key

ZVPDNRVYHLRXLX-UHFFFAOYSA-N

General description

A cell-permeable pyrazolopyrimidine compound that is shown to inhibit Src family tyrosine kinases Lck, Fyn, Hck, and Src (IC50 = 5, 6, 20, and 170 nM, respectively), as well as non-Src family kinases such as CSK, RIP2, and CK1δ (IC50 = 640 nM, 26 nM, 170 nM, respectively), in in vitro kinase assays. At 1 µM, in vitro kinase activity is reduced by more than 57% for p38α, p38β, CK1δ, CSK, RIP2, and GAK, while exhibiting much lower activity against more than 60 other kinases. Kinetic studies employing Src and [poly Glu-Tyr 4:1] indicate that the inhibition is mixed competitive with respect to the substrate and non-competitive with respect to ATP. PP1 also demonstrates anti-growth properties in several B lymphoma cultures of human and murine origins (IC50 = 2.5 to 8.0 µM) and pediatric brain tumor cells (IC50 = 1.1 to 9.7 µM). Also available in InSolution format (Cat. No. 539741.
A cell-permeable pyrazolopyrimidine compound that is shown to inhibit Src family tyrosine kinases Lck, Fyn, Hck, and Src (IC50 = 5, 6, 20, and 170 nM, respectively), as well as non-Src family kinases such as CSK, RIP2, and CK1δ (IC50 = 640 nM, 26 nM, 170 nM, respectively), in in vitro kinase assays. At 1 µM, in vitro kinase activity is reduced by more than 57% for p38α, p38β, CK1δ, CSK, RIP2, and GAK, while exhibiting much lower activity against more than 60 other kinases. Kinetic studies employing Src and [poly Glu-Tyr 4:1] indicate that the inhibition is mixed competitive with respect to the substrate and non-competitive with respect to ATP. PP1 also demonstrates anti-growth properties in several B lymphoma cultures of human and murine origins (IC50 = 2.5 to 8.0 µM) and pediatric brain tumor cells (IC50 = 1.1 to 9.7 µM). Also available in InSolution format (Cat. No. 539741).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ke, J., et.al. 2009. Mol. Cancer8, 132.
Sikkema, A.H., et al. 2009. Cancer Res.69, 5987.
Bain, J., et al. 2007. Biochem. J.408, 297.
Bain, J., et al. 2003. Biochem. J.371, 199.
Karni, R., et al. 2003. FEBS Lett.537, 47.
Hanke, J.H., et al. 1996. J. Biol. Chem.271, 695.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service