Skip to Content
Merck
All Photos(1)

Key Documents

1.16899

Supelco

Ammonium Test Strips, reflectometric

5.0-20.0 mg/L (NH₄⁺), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Ammonium chemical test strips, Ammonium test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Ammonium Test, reflectometric, 5.0-20.0 mg/L (NH4+), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

ammonium

measuring range

5.0-20.0 mg/L (NH4+)

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: Reflectometric with test strips and reagents, 5.0 - 20.0 mg/L (NH4+),Reflectoquant®

The Reflectoquant® Ammonium test includes test strips and all necessary additional reagents and is suitable for determination potentially harmful pollution in surface and ground water, which are caused by ammonium compounds. Depending on the pH level ammonia is formed, which is a potent toxin for fish. In agriculture the Reflectoquant® test strips can be used to measure the ammonium nitrogen content of various types of manure and compost after sample preparation. Ammonium ions react with a chlorinating agent to form monochloramine. This in turn reacts with a phenol compound to form a blue indophenol derivative that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the ammonium concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant® test strips for ammonium. Further quantitative determination methods can be found with our Spectroquant® tests.

Application


  • Development of celiac-safe foods: Explores the role of zinc sulfate in preventing gluten deamidation, an innovative approach to producing celiac-safe foods, showcasing its utility in food science and human health (Engström et al., 2024).



Analysis Note

function control: conforms
Assay
- standard 1:
6.5 - 8.5 mg/l
- standard 2: 9.0 - 13.0 mg/l
- standard 3: 13.5 - 17.5 mg/l

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Storage Class Code

8B - Non-combustible, corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Step-by-step accurate reflectometric determination of ammonium in wastewater with Nessler’s reagent or Indophenol blue using Reflectroquant®system and test strips.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service