Skip to Content
Merck
All Photos(1)

Key Documents

444234

Sigma-Aldrich

MMP-9-Lipocalin-TIMP-1 Complex, Human Neutrophil

Synonym(s):

Matrix Metalloproteinase 9-Lipocalin-TIMP-1 Complex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥600 mU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

other MMP activity, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Native MMP-9-Lipocalin-TIMP-1 complex from stimulated human neutrophils. The ternary MMP-9/lipocalin/TIMP-1 complex behaves like the progelatinase B/TIMP-1 complex in acting as an inhibitor of active MMPs. Following activation, the complex acts as a gelatinase with reduced activity.
Note: 1 mU = 1 milliunit.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol 2,4-DNP-Pro-Gln-Gly-Ile-Ala-Gly-Gln-D-Arg-OH per min at 37°C, pH 7.0.

Physical form

In 200 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0.05% Brij® 35 Detergent, 0.05% NaN₃, pH 7.0.

Preparation Note

Prepared from stimulated neutrophils that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kolkenbrock, H., et al. 1996. Biol. Chem. 377, 529.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service