Z742919
Detergent 8®, liquid detergent
pkg of 19 L (Jerrycan)
Synonym(s):
biodegradable detergent, non-ionic detergent, ultrasonic cleaner detergent
Select a Size
$806.00
Select a Size
About This Item
$806.00
Recommended Products
description
Biodegradable
packaging
pkg of 19 L (Jerrycan)
manufacturer/tradename
Alconox, Inc. 1705
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Used to clean: Circuit boards, electronic parts, phosphate sensitive labware, nuclear reactor cavities, delicate industrial parts and nuclear contaminated equipment during outage. FDA certified.
Used to remove: Resins, rosins, oils, dirt, grime, particulates, residues, fluxes, chemicals and solvents.
Surfaces cleaned: Corrosion inhibited formulation recommended for glass, metal, stainless steel, ceramic, plastic, and circuit boards. Can be used, with prompt rinsing, on soft metals such as copper, aluminum, silver, and gold. Aluminum can darken and mixed metal galvanic reactions can occur. Corrosion testing is advisable.
Cleaning method: Soak, brush, sponge, cloth, ultrasonic, ware washer, spray washer, pressure washer and flow through clean-in-place.
Directions: For manual, soak and ultrasonic use, make a 3-5% solution (4-6 oz. per gal. or 30-50 ml per liter) in warm or hot water. An oily, milky solution is normal.
Protective gloves and eyewear are recommended.
For spray machines, add detergent at the hot water wash cycle to make a 2% solution (2 1/2 oz. per gal. or 20 ml per liter). For difficult soils, raise water temperature and use more detergent. RINSE
THOROUGHLY—preferably with running water.
For critical cleaning, do final or all rinsing in distilled, deionized or purified water. Used on a range of glass, ceramic, plastic and metal surfaces. Corrosion testing is advisable.
Features and Benefits
- Biodegradable; contains no harmful phosphates, making it environmentally responsible.
- Low-foaming and ion-free formulation for cleaning delicate industrial parts without residue.
- Time-saving with strong wetting and penetrating power, enhancing cleaning efficiency.
- Effective at removing resins, oils, dirt, and grime from a wide range of surfaces.
- Certified by the FDA, ensuring compliance with safety and regulatory standards.
- Compatible with soft metals like aluminum and silver when corrosion testing is followed.
- Suitable for both manual and automated cleaning systems, offering operational flexibility.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
159.8 °F
Flash Point(C)
71 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service