Skip to Content
Merck
All Photos(2)

Documents

1.71200

Supelco

Thermoreactor

number of holes 12 hole, Spectroquant® TR 320

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
SE.11

manufacturer/tradename

Spectroquant® TR 320

technique(s)

photometry: suitable

storage temp.

2-30°C

General description

12 holes, 7 preinstalled programmes: 148 °C (20, 120 min), 150 °C (120 min), 120 °C (30, 60, 120 min), 100 °C (60 min) TR 320 Spectroquant®
The Spectroquant Thermoreactor TR 320 is designed for the digestion up to 12 samples simultaneously. Precisely controlled thermal digestion is a key sample preparation step during the determination of COD, TOC, total contents of cadmium, chromium, cyanide, iron, lead, nickel, nitrogen, phosphorus, silver, or zinc using our high quality Spectroquant Test Kits.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination of total nitrogen in soil samples by phosphorus molybdenum blue method using Spectroquant® test kits.

Cobalt in wastewater from the production and casting of non-ferrous metals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service