Skip to Content
Merck
All Photos(1)

Key Documents

1.01701

Millipore

N⁶-Benzyladenine

≥99.0% (HPLC)

Synonym(s):

N⁶-Benzyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
MDL number:
UNSPSC Code:
12352116
EC Index Number:
214-927-5

Assay

≥99.0% (HPLC)

InChI

1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)

InChI key

NWBJYWHLCVSVIJ-UHFFFAOYSA-N

General description

for biochemistry

Analysis Note

Assay (HPLC): ≥ 99 %
Identity (IR-spectrum): passes test
Absorption maximum (buffer pH 2.0): 271 - 278 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.0005 %; buffer pH 2.0): 830 - 870

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service