Skip to Content
Merck
All Photos(2)

Key Documents

1.05886

Supelco

Magnesium sulfate heptahydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

bitter salt, epsom salt, sulfuric acid magnesium salt heptahydrate, Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-298-2
NACRES:
NA.21
Assay:
≥99.5% (complexometric)
99.0-100.5% dry basis (complexometric)
grade:
ACS reagent
form:
solid
solubility:
710 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (complexometric)
99.0-100.5% dry basis (complexometric)

form

solid

impurities

≤0.005% Insoluble matter

loss

48.0-52.0% loss on drying, 400 °C

pH

5.0-8.0 (25 °C, 50 g/L in H2O)

solubility

710 g/L

density

1.68 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.0003%
nitrate (NO3-): ≤0.002%

cation traces

As: ≤0.0002%
Ca: ≤0.005%
Cu: ≤0.0001%
Fe: ≤0.0001%
K: ≤0.001%
Mn: ≤0.0005%
N: ≤0.002%
NH4+: ≤0.002%
Na: ≤0.001%
Pb: ≤0.0001%
Sr: ≤0.005%
heavy metals: ≤0.0005%

storage temp.

2-30°C

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Bioflocculant produced by Bacillus velezensis and its potential application in brewery wastewater treatment.: This research explores the application of a bioflocculant produced by Bacillus velezensis, with magnesium sulfate heptahydrate as a potential flocculating agent in brewery wastewater treatment. The study highlights the efficacy of magnesium sulfate heptahydrate in enhancing flocculant performance, offering a cost-effective and eco-friendly solution for wastewater management in the beverage industry (Agunbiade et al., 2022).

Analysis Note

Assay (complexometric): ≥ 99.5 %
Assay (complexometric; calculated on dried substance): 99.0 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.005 %
Acidity or alkalinity: passes test
pH-value (5 %; water; 25 °C): 5.0 - 8.0
Chloride (Cl): ≤ 0.0003 %
Total nitrogen (N): ≤ 0.002 %
Nitrate (NO₃): ≤ 0.002 %
Heavy metals (ACS): ≤ 0.0005 %
As (Arsenic): ≤ 0.0002 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.0001 %
K (Potassium): ≤ 0.001 %
Mn (Manganese): ≤ 0.0005 %
Na (Sodium): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.002 %
Pb (Lead): ≤ 0.0001 %
Sr (Strontium): ≤ 0.005 %
Loss on drying (400 °C): 48.0 - 52.0 %
Corresponds to ACS, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service