Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
1.00955
1,2-Dichloroethane
EMPLURA®
Synonym(s):
Ethylene chloride, Ethylene dichloride
About This Item
Recommended Products
vapor density
3.4 (20 °C, vs air)
Quality Level
vapor pressure
87 mmHg ( 25 °C)
product line
EMPLURA®
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
775 °F
potency
670 mg/kg LD50, oral (Rat)
2800 mg/kg LD50, skin (Rabbit)
expl. lim.
15.6 %
dilution
(low-cost alternative, for basic applications)
impurities
≤0.000002% Cd (Cadmium)
≤0.000002% Co (Cobalt)
≤0.000002% Cr (Chromium)
≤0.000002% Cu (Copper)
≤0.000002% Mn (Manganese)
≤0.000002% Ni (Nickel)
≤0.000002% Pb (Lead)
≤0.000002% Sr (Strontium)
≤0.000005% Al (Aluminium)
≤0.000005% Ba (Barium)
≤0.000005% Mg (Magnesium)
≤0.00001% Zn (Zinc)
≤0.00002% Ca (Calcium)
≤0.00002% K (Potassium)
≤0.00003% Free chlorine (Cl)
≤0.00005% Na (Sodium)
≤0.0001% Fe (Iron)
≤0.0003 meq/g Titrable acid
≤0.030% Water
color
APHA: ≤50
refractive index
n20/D 1.444 (lit.)
bp
83 °C (lit.)
mp
−35 °C (lit.)
transition temp
flash point 13 °C
density
1.256 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
ClCCCl
InChI
1S/C2H4Cl2/c3-1-2-4/h1-2H2
InChI key
WSLDOOZREJYCGB-UHFFFAOYSA-N
Related Categories
General description
Specifications
Analysis Note
≤ 0.030 %≤ 0.030 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
55.4 °F - closed cup
Flash Point(C)
13 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service