Skip to Content
Merck
All Photos(1)

Key Documents

DX0800

Supelco

1,2-Dichloroethane

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Dichloroethane, Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-458-1
grade:
reagent
bp:
83 °C (lit.)
vapor pressure:
87 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

775 °F

expl. lim.

15.6 %

dilution

(for analytical testing)

impurities

≤0.0003 meq/g Titrable acid
≤0.03% Water (H2O)

evapn. residue

≤0.002%

color

clearAPHA: ≤10

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

transition temp

flash point 13 °C

solubility

7.9 g/L

density

1.25 g/cm3 at 20  °C
1.256 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Green Catalytic Synthesis of Ethylenediamine from Ethylene Glycol and Monoethanolamine: A Review: This review details a green synthesis method using 1,2-dichloroethane, enhancing the sustainability of chemical processes in academia and industrial applications (Wu and An, 2024).
  • The complete degradation of 1,2-dichloroethane in Escherichia coli by metabolic engineering: This study demonstrates a biotechnological approach to degrade 1,2-dichloroethane, reducing environmental impact, relevant for both academia and biotechnological industries (Deng et al., 2024).
  • Characteristics of Volatile Organic Compounds Emitted from Airport Sources and Their Effects on Ozone Production: Investigates the role of 1,2-dichloroethane among other VOCs in environmental pollution and ozone formation, crucial for environmental research and regulations (Chen et al., 2024).

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Appearance: Clear
Assay (GC): 99.0% min
Color (APHA): 10 max
Evaporation residue: 0.002% max
Form: Liquid
Titrable acid: 0.0003meq/g max
Water (H2O): 0.03% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service