Skip to Content
Merck
All Photos(1)

Key Documents

1.00087

Supelco

Chlorine Test, photometric

photometric, 0.010 - 6.00 mg/l Cl₂ for free chlorine: Cl₂-1 and Cl₂-2, for total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Chlorine reagent Cl₂-2 (liquid), photometric, 0.010 - 6.00 mg/l Cl₂ for free chlorine: Cl₂-1 and Cl₂-2, for total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®

Quality Level

form

liquid

usage

sufficient for 400 tests

specific analyte(s)

chlorine

measuring range

0.010-6.00 mg/L (Cl2)

technique(s)

photometry: suitable

density

1.079 g/cm3 at 20 °C

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Move DC
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (DPD)

storage temp.

15-25°C

General description

We offer Spectroquant® bulk reagents for the determination of free and total chlorine. This article contains reagent Cl2-2 . For the determination the following articles have to be purchased in addition: Reagents 1.00086, 1.00088 (only for the determination of total chlorine) and the equipment 1.00089. More information can be found in the instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for total chlorine.

AQA Standard for free chlorine - Preparation of a standard solution for free chlorine

Preparation of a standard solution for free Chlorine according to DIN EN ISO 7393

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service