Skip to Content
Merck
All Photos(1)

Documents

E-091

Supelco

Eutylone-D5 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12D5NO3 · HCl
Molecular Weight:
276.77
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C(CC)NC([2H])([2H])C([2H])([2H])[2H])C1=CC(OCO2)=C2C=C1.[H]Cl

InChI

1S/C13H17NO3.ClH/c1-3-10(14-4-2)13(15)9-5-6-11-12(7-9)17-8-16-11;/h5-7,10,14H,3-4,8H2,1-2H3;1H/i2D3,4D2;

InChI key

LZTAJXOEHCOKDE-CIIWIYAOSA-N

General description

Eutylone is a synthetic cathinone found in numerous bath salt blends under the street name bk-MBDB. This designer drug and stimulant is abused in the US and Europe. This stable-labeled internal standard is suitable for quantification of eutylone using LC/MS or GC/MS methods for applications in clinical toxicology, forensic analysis, and urine drug testing applications.

Application


  • Eutylone-D₅ hydrochloride as a research chemical: Eutylone-D₅ hydrochloride is extensively utilized as a research chemical for analytical research, providing a stable isotope-labeled standard that facilitates accurate measurements in mass spectrometry, particularly in studies involving psychoactive substances and their effects on serotonin uptake (AlSalem et al., 2024).

  • Stable isotope-labeled Eutylone for pharmacokinetic studies: This compound is crucial in pharmacokinetic studies to understand the drug′s absorption, distribution, metabolism, and excretion, aiding researchers in developing safer and more effective therapeutic interventions (Deshpande et al., 2024).

  • Internal standard for mass spectrometry in biochemical research: Eutylone-D₅ hydrochloride serves as a reliable internal standard in mass spectrometry, ensuring precision in the quantitative analysis of complex biological samples and in the study of biochemical pathways related to psychoactive substances (Elawadi et al., 2024).

  • High-purity Eutylone-D₅ HCl for drug metabolism research: The high purity of Eutylone-D₅ hydrochloride makes it an essential tool in drug metabolism research, helping to identify metabolites and understand the metabolic fate of synthetic cannabinoids (Durgun et al., 2024).

  • Forensic toxicology analysis: In forensic toxicology, Eutylone-D₅ hydrochloride is used to detect and quantify the presence of Eutylone in biological samples, which is critical in legal and clinical cases involving drug use (Puvvula et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service