E-087
Eutylone hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Narcotic Licence Schedule E (Switzerland)
concentration
1.0 mg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
N(C(CC)C(=O)c1cc2c(cc1)OCO2)CC
InChI
1S/C13H17NO3/c1-3-10(14-4-2)13(15)9-5-6-11-12(7-9)17-8-16-11/h5-7,10,14H,3-4,8H2,1-2H3
InChI key
YERSNXHEOIYEGX-UHFFFAOYSA-N
General description
Application
Eutylone hydrochloride, a powerful psychoactive substance, is extensively employed in neuropharmacology research to explore the impact of synthetic cathinones on neurotransmitter systems. Its use facilitates the examination of intricate mechanisms underlying stimulant abuse and addiction, offering valuable insights into potential therapeutic approaches for treating drug addiction ( 1).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F
Flash Point(C)
9.7 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service