Skip to Content
Merck
All Photos(1)

Key Documents

878122P

Avanti

C16-20:3 PC

Avanti Research - A Croda Brand 878122P, powder

Synonym(s):

1-O-hexadecyl-2-(8Z,11Z,14Z-eicosatrienoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$710.00

$710.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$710.00

About This Item

Empirical Formula (Hill Notation):
C44H84NO7P
CAS Number:
Molecular Weight:
770.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$710.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (878122P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878122P

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCC/C=C\C/C=C\C/C=C\CCCCC)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C44H84NO7P/c1-6-8-10-12-14-16-18-20-22-23-24-25-27-29-31-33-35-37-44(46)52-43(42-51-53(47,48)50-40-38-45(3,4)5)41-49-39-36-34-32-30-28-26-21-19-17-15-13-11-9-7-2/h14,16,20,22,24-25,43H,6-13,15,17-19,21,23,26-42H2,1-5H3/b16-14-,22-20-,25-24-/t43-/m1/s1

InChI key

VYKSGWCIVDQIQR-MEAIPJDCSA-N

General description

Phophotidylcholine (PC) is a major phospholipid present abundantly in eukaryotic membranes. It is originated from the Kennedy (CDP‐choline) pathway.[1]

Biochem/physiol Actions

Phophotidylcholine (PC) is involved in maintaining the integrity of membranes. Increase in levels of PC is associated with increase in cell growth. It is necessary for the myelination of nerve cells and production of very low density lipoprotein (VLDL) from hepatocytes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (878122P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From yeast to humans-roles of the Kennedy pathway for phosphatidylcholine synthesis
McMaster C R
Febs Letters, 592(8), 1256-1272 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service