Skip to Content
Merck
All Photos(2)

Key Documents

852469C

Avanti

C18(Plasm)-20:4 PC

1-(1Z-octadecenyl)-2-arachidonoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-O-1′-(Z)-octadecenyl-2--(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphocholine; GPCho(18:0p/20:4(5Z,8Z,11Z,14Z))

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$436.00

$436.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$436.00

About This Item

Empirical Formula (Hill Notation):
C46H84NO7P
CAS Number:
Molecular Weight:
794.14
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$436.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mL (852469C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 852469C

concentration

1 mg/mL (852469C-5mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

Application

C18(Plasm)-20:4 PC has been used:
  • as a lipid standard in liquid chromatography-mass spectrometry (LC-MS) method[1]
  • in the preparation of large unilamellar vesicles (LUVs)[2]
  • for determining the substrate specificity of glycerophosphocholine cholinephosphodiesterase-14 (GPC-CP14) protein from Streptomyces sanglieri [3]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (852469C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Sugimori et al.
Journal of bioscience and bioengineering, 117(4), 422-430 (2013-11-12)
A novel metal ion-independent glycerophosphocholine cholinephosphodiesterase (GPC-CP) of Streptomyces sanglieri was purified 53-fold from culture supernatant with 1.1% recovery (583 U/mg-protein). The enzyme functions as a monomer with a molecular mass of 66 kDa. The gene encoding the enzyme consists
Comprehensive untargeted lipidomic analysis using core-shell C30 particle column and high field orbitrap mass spectrometer
Narvaez-Rivas M and Zhang Q
Journal of Chromatography A, 1440, 123-134 (2016)
Cytochrome c is an oxidative stress-activated plasmalogenase that cleaves plasmenylcholine and plasmenylethanolamine at the sn-1 vinyl ether linkage
Jenkins CM, et al.
The Journal of Biological Chemistry, 293(22), 8693-8709 (2018)
Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service