Skip to Content
Merck
All Photos(2)

Key Documents

850458P

Avanti

16:0-18:2 PC

1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-hexadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphocholine; PC(16:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$375.00
200 MG
$1,220.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$375.00
200 MG
$1,220.00

About This Item

Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$375.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (850458P-200mg)
pkg of 1 × 25 mg (850458P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850458P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

General description

16:0-18:2 PC or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphocholine is a glycerophospholipid, with substitution of palmitic acid and linolenic acid at sn-1 and sn-2 positions of the glycerol backbone, respectively.

Application

16:0-18:2 PC or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphocholine:
  • has been used in liposomes for standard cell-free translation[1]
  • has been used in the extraction of serum lipids and in peroxisome proliferator-activated receptor-δ (PPAR-δ) assay[2]
  • might be used to study the effects of phosphatidylcholines (PCs) on lubrication of damaged mesothelium[3]

Packaging

5 mL Clear Glass Sealed Ampule (850458P-200mg)
5 mL Clear Glass Sealed Ampule (850458P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lindsay Kosack et al.
Scientific reports, 7(1), 11289-11289 (2017-09-14)
Lipid metabolism is increasingly being appreciated to affect immunoregulation, inflammation and pathology. In this study we found that mice infected with lymphocytic choriomeningitis virus (LCMV) exhibit global perturbations of circulating serum lipids. Mice lacking the lipid-sensing surface receptor triggering receptor
Giulia Coliva et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-25)
Free radical driven lipid peroxidation is a chain reaction which can lead to oxidative degradation of biological membranes. Propagation vs. termination rates of peroxidation in biological membranes are determined by a variety of factors including fatty acyl chain composition, presence
Francesca Bodega et al.
Respiratory physiology & neurobiology, 203, 116-120 (2014-08-17)
Effect of time and phosphatidylcholines (PCs) on lubrication of damaged mesothelium has been investigated. Marked increase in coefficient of kinetic friction (μ) of pleural specimens after mesothelial blotting and rewetting decreased by 23.4±3.5%, 41.8±3.8%, and 40.5±2.7% after 30min, 1h, and
Eikan Mishima et al.
Journal of the American Society of Nephrology : JASN, 31(2), 280-296 (2019-11-27)
Ferroptosis, nonapoptotic cell death mediated by free radical reactions and driven by the oxidative degradation of lipids, is a therapeutic target because of its role in organ damage, including AKI. Ferroptosis-causing radicals that are targeted by ferroptosis suppressors have not
John K Eaton et al.
Nature chemical biology, 16(5), 497-506 (2020-04-02)
We recently described glutathione peroxidase 4 (GPX4) as a promising target for killing therapy-resistant cancer cells via ferroptosis. The onset of therapy resistance by multiple types of treatment results in a stable cell state marked by high levels of polyunsaturated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service