Skip to Content
Merck
All Photos(2)

Key Documents

852804C

Avanti

C18(Plasm)-20:4 PE

1-(1Z-octadecenyl)-2-arachidonoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1-O-1′-(Z)-octadecenyl-2--(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphoethanolamine; GPEtn(18:0p/20:4(5Z,8Z,11Z,14Z))

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$861.00

$861.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$861.00

About This Item

Empirical Formula (Hill Notation):
C43H78NO7P
CAS Number:
Molecular Weight:
752.06
UNSPSC Code:
51191904
NACRES:
NA.25

$861.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (852804C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 852804C

concentration

1 mg/mL (852804C-5mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[NH3+])(OC[C@]([H])(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)CO/C=C\CCCCCCCCCCCCCCCC)=O

InChI

1S/C43H78NO7P/c1-3-5-7-9-11-13-15-17-19-21-22-24-26-28-30-32-34-36-43(45)51-42(41-50-52(46,47)49-39-37-44)40-48-38-35-33-31-29-27-25-23-20-18-16-14-12-10-8-6-4-2/h11,13,17,19,22,24,28,30,35,38,42H,3-10,12,14-16,18,20-21,23,25-27,29,31-34,36-37,39-41,44H2,

InChI key

URPXXNCTXCOATD-FXMFQVEGSA-N

General description

C18(Plasm)-20:4 PE are plasmenylethanolamines that contains arachidonic acid. Plasmalogens, in general, contain a vinyl-ether linkage at the sn-1 position and at sn-2 position it is highly enriched with polyunsaturated fattyacid, specifically, arachidonic acid (AA).[1]

Application

C18(Plasm)-20:4 PE has been used in the preparation of 2-arachidonic acid-lysophosphatidylethanolamine (2-AA-LPE).[1] It has also been used in the synthesis of biotinylated analyte standard for reverse phase-high performance liquid chromatography (HPLC)-electrospray ionization-mass spectroscopy.[2]

Biochem/physiol Actions

Plasmalogens are phospholipids, essential for cell function and signaling. Plasmalogens influences membrane structure and molecular dynamics, contributes to the functioning of integral membrane proteins. It plays an important role in the release of membrane fatty acid for signaling upon cell stimulation.[1]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (852804C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and quantification of aminophospholipid molecular species on the surface of apoptotic and activated cells
Thomas CP, et al.
Nature Protocols, 9(1), 51-51 (2014)
Cytochrome c is an oxidative stress-activated plasmalogenase that cleaves plasmenylcholine and plasmenylethanolamine at the sn-1 vinyl ether linkage
Jenkins CM, et al.
The Journal of Biological Chemistry, 293(22), 8693-8709 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service