Skip to Content
Merck
All Photos(2)

Key Documents

850544O

Avanti

10MAG

Avanti Research - A Croda Brand 850544O

Synonym(s):

1-decanoyl-rac-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$243.00

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$243.00

About This Item

Empirical Formula (Hill Notation):
C13H26O4
CAS Number:
Molecular Weight:
246.34
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

mol wt

246.34 g/mol

packaging

pkg of 10 × 0.1 g (with screw cap (850544O-1g))

manufacturer/tradename

Avanti Research - A Croda Brand 850544O

technique(s)

NMR: suitable
bio NMR: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(OCC(O)CO)CCCCCCCCC

InChI

1S/C23H44O5/c1-3-5-7-9-11-13-15-17-22(25)27-20-21(19-24)28-23(26)18-16-14-12-10-8-6-4-2/h21,24H,3-20H2,1-2H3

InChI key

GNSDEDOVXZDMKM-UHFFFAOYSA-N

General description

An optimized reverse micelle surfactant system has been developed for solution nuclear magnetic resonance studies of encapsulated proteins and nucleic acids dissolved in low viscosity fluids. Comprising the nonionic 1-decanoyl-rac-glycerol and the zwitterionic lauryldimethylamine-N-oxide (10MAG/LDAO), this mixture is shown to efficiently encapsulate a diverse set of proteins and nucleic acids. Chemical shift analyses of these systems show that high structural fidelity is achieved upon encapsulation.
Monoglycerides/monoacylglycerols are polar lipids and are amphiphilic in nature.[1]10MAG (1-decanoyl-rac-glycerol) is a nonionic surfactant.[2]

Application

10MAG (1-decanoyl-rac-glycerol) may be used:
  • as a substrate for fluorescent glycerol assay[3]
  • as a standard for the quantification of monoacylglycerols (MAGs) in micellar fraction analysis[4]
  • as a component in the surfactant mixture for reverse micelle sample preparation[2]

Biochem/physiol Actions

Monoglycerides are major component of baking mixtures.[5]

Packaging

5 mL PTFE Vial with Screw Cap (850544O-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen G Valentine et al.
Journal of the American Chemical Society, 136(7), 2800-2807 (2014-01-25)
Despite tremendous advances in recent years, solution NMR remains fundamentally restricted due to its inherent insensitivity. Dynamic nuclear polarization (DNP) potentially offers significant improvements in this respect. The basic DNP strategy is to irradiate the EPR transitions of a stable
Fat as, or in, food
Fats and oils handbook, 719-802 null
Igor Dodevski et al.
Journal of the American Chemical Society, 136(9), 3465-3474 (2014-02-06)
An optimized reverse micelle surfactant system has been developed for solution nuclear magnetic resonance studies of encapsulated proteins and nucleic acids dissolved in low viscosity fluids. Comprising the nonionic 1-decanoyl-rac-glycerol and the zwitterionic lauryldimethylamine-N-oxide (10MAG/LDAO), this mixture is shown to
Heloísa Helena de Abreu-Martins et al.
Food chemistry, 311, 126024-126024 (2019-12-20)
Solid lipid nanoparticles (SLNs) are emulsion-based carriers of lipophilic bioactive compounds. However, their digestibility may be affected by the solid lipid phase composition. Hence, the aim of this work was to study the in vitro lipolysis kinetics as well as
Juha R Savinainen et al.
PloS one, 9(10), e109869-e109869 (2014-10-08)
Human lymphocyte antigen B-associated transcript 5 (BAT5, also known as ABHD16A) is a poorly characterized 63 kDa protein belonging to the α/β-hydrolase domain (ABHD) containing family of metabolic serine hydrolases. Its natural substrates and biochemical properties are unknown. Amino acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service